Drying tubes, calcium chloride
Leverandør: KARTELL
446 447 448
468-1210EA 155 NOK
468-1210 468-1211 468-1212
Drying tubes, calcium chloride
Rør Tørkerør
PE.
  • Both ends have tapered, push-on connectors
Suitable for drying gases.