Reichardt′s dye

Leverandør: Thermo Fisher Scientific

Synonymer: 5'-(2,4,6-Triphenylpyridin-1-ium-1-yl)-[1,1':3',1''-terphenyl]-2'-olate, 2,6-Diphenyl-4-(2,4,6-triphenyl-1-pyridinio)phenolate, 2,6-Diphenyl-4-(2,4,6-triphenylpyridinio)phenolate

468370010
468370010.EA 4050 NOK
468370010.
Reichardt′s dye
Reichardt′s dye
Formel: C₄₁H₂₉NO
MW: 551.68 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013355
CAS-nummer: 10081-39-7

Bestill nå

Spesifikasjon Testresultater

Appearance dark blue to black powder
Infrared spectrum Conforms
UV Lambda max 304 to 308 nm
Specific extinction E (1%/1cm) ≥35000 (0.008 g/l in methanol)
Elemental Analysis 79.4 to 99.1 % (C) / 2.1 to 2.8 (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR