Cediranib maleate 98%

Leverandør: Thermo Fisher Scientific

468952500
468952500.EA 6150 NOK
468952500.
Cediranib maleate 98%
Cediranib maleate

Bestill nå

Spesifikasjon Testresultater

Appearance white to beige powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with NaOH 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR