Kobber ≥99.8% (metals basis), folie, annealed, uncoated, Thickness: 0.025 mm (0.001 in)

Leverandør: Thermo Fisher Scientific

46986.RH 46986.RF 46986.RG
46986.RHEA 8800 NOK
46986.RH 46986.RF 46986.RG
Kobber ≥99.8% (metals basis), folie, annealed, uncoated, Thickness: 0.025 mm (0.001 in)
Kobber

Purity: 99.8% CAS No.: 7440-50-8 Physical Form: ≈20.2g/30x30cm Appearance: Copper Odour: Odourless MDL No.: MFCD00010965

Formel: Cu
Kokepunkt: 2595 °C (1013 hPa)
Smeltepunkt: 1083 °C
Tetthet: 8,94 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010965
CAS-nummer: 7440-50-8
EINECS: 231-159-6
Merck Indeks: 13,02545

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR