Svovelsyre concentrate for dilution to 1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®

Leverandør: Avantor
Danger

4700.EA 415 NOK
4700.
Svovelsyre concentrate for dilution to 1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Svovelsyre

(1 equiv.=49.04 g H2SO4)

Formel: H₂SO₄
MW: 98.08 g/mol
Kokepunkt: 330
Smeltepunkt: 10.38
Tetthet: 1.84
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064589
CAS-nummer: 7664-93-9
EINECS: 231-639-5
FN: 2796
ADR: 8 II

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use Passes Test
Normality (by titrimetry) Passes Test
(Makes 1N solution after dilution to 1000 mL)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR