Laboratory refrigerator, LAB300
Leverandør: PORKKA SCANDINAVIA
LAB300
471-0740EA 0 NOK
471-0740
Laboratory refrigerator, LAB300
Kjøleskap
Designed and manufactured for the exacting standards of biological and pharmaceutical laboratories as well as Life Sciences.
  • Spark proof interior, seven stainless steel plate shelves
  • Digital temperature display with built-in microprocessor controller
  • Visual and audible high/low temperature alarm; recording of alarms with date and time
  • Display temperature accuracy down to 1 °C
  • Energy efficient and environmentally friendly (HFC free polyurethane) insulation material
  • Door equipped with lock and magnetic gasket
Exterior finish in white polyester-coatedMore Product Information