Ultra-Low Temperature Freezers, TSX
Leverandør: Thermo Scientific
TSX60086V
471-1205EA 0 NOK
471-1205
Ultra-Low Temperature Freezers, TSX
Frysere Frysere, ultra-
Thermo Scientific™ TSX series ultra-low high performance freezers are designed with features that support sustainability objectives without compromising performance. The series' V-drive technology is designed to provide temperature uniformity that adapts to the laboratory environment, offering significant energy savings. Now available in four capacities, these −80 °C freezers maximise sample storage capacity and offer substantial energy savings over conventional refrigerant ultra-low freezers.
  • Best peak variation of energy efficient, ultra-low freezers: Less than 5 °C
  • Record