Ultra-low temperature upright freezers, model SU780XLE
Leverandør: Stirling Ultracold
SU780XLE-EU
471-1238EA 0 NOK
471-1238
Ultra-low temperature upright freezers, model SU780XLE
Frysere Frysere, ultra-
Ultra-low temperature freezer featuring the free-piston Stirling engine. This next generation of environmentally sustainable ULT freezers achieve stable storage conditions over a wide temperature range. The freezer uses 70 to 75% less energy than standard compressor-based ULT freezers, with the industry's best ultra-low cooling performance.
  • ±1 °C 'steady state' temperature variation over time
  • Faster initial pull down, ambient to –80 °C, <6,5 hours
  • Faster door opening temperature recovery, 35 minutes to –80 °C (ENERGY STAR® final test method)
  • Slowest