Precision digital thermometer (NTC and Pt100) with app connection, testo 110
Leverandør: Testo
0563.0110
471-1335EA 1850 NOK
471-1335
Precision digital thermometer (NTC and Pt100) with app connection, testo 110
Termometere Digitaltermometer
The compact temperature measuring instrument testo 110 convinces every day in use, shows its strengths wherever measurements are to be made at particularly low temperatures or where highly precise temperatures are to be determined.
The fast temperature all-rounder can show its strengths particularly in applications in laboratories or in the food environment: The optionally available NTC or Pt100 probes cover a comprehensive range of measuring tasks and ensure exact results with particularly high accuracy. The digital Pt100 probes can also be calibrated without the measuring instrument, soMore Product Information