Antimony, plasma standard solution, Sb 1000 µg/ml, Specpure®

Leverandør: Thermo Fisher Scientific

47103.ADEA 925 NOK
47103.AD 47103.AE
Antimony, plasma standard solution, Sb 1000 µg/ml, Specpure®
Standarder Kationstandarder Antimony Standards

Molecular Formula: Sb in 30% HCl
Physical Form: Liquid
Odour: Acidic
UN No.: UN1789
MDL No.: MFCD00134030

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR