Stivelse, løselig 1% (w/v) vandig løsning for iodometric titrations

Leverandør: Thermo Fisher Scientific

47209.AP 47209.K2 47209.K7 47209.M4
47209.APEA 485 NOK
47209.AP 47209.K2 47209.K7 47209.M4
Stivelse, løselig 1% (w/v) vandig løsning for iodometric titrations
Stivelse, løselig

CAS No.: 9005-84-9 Physical Form: Liquid for iodometric titrations MDL No.: MFCD00082026

Formel: (C₆H₁₀O₅)n
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00082026
CAS-nummer: 9005-84-9
EINECS: 232-686-4
Merck Indeks: 14,8799

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR