Intermediate polar column test mix, certified reference material, Supelco®
Leverandør: Merck
47301
47301.EA 645 NOK
47301.
Intermediate polar column test mix, certified reference material, Supelco®
Standarder PetroleumIndustristandarder
Analytes:
Decane; 2,6-Dimethylaniline; 2,6-Dimethylphenol; Dodecane; 1-Octanol; 2-Octanone; Tetradecane; Tridecane; Undecane
Sertifiseringer: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.