Programmed test mix, in dichloromethane (varied conc.), certified reference material, Supelco®
Leverandør: Merck
47304
47304.EA 750 NOK
47304.
Programmed test mix, in dichloromethane (varied conc.), certified reference material, Supelco®
Standarder kromatografi standarder
This test mix is based on the comprehensive mix developed by Grob (Grob, et al., J. Chromatogr. 156 (1978) p. 1)
Analytes:
2,3-Butanediol 530 μg/ml; Decane 280 μg/ml; Dicyclohexylamine 310 μg/ml; 2,6-Dimethylaniline 320 μg/ml; 2,6-Dimethylphenol 320 μg/ml; 2-Ethylhexanoic acid 380 μg/ml; Methyl decanoate 420 μg/ml; Methyl laurate 410 μg/ml; Methyl undecanoate 420 μg/ml; Nonanal 400 μg/ml; 1-Octanol 360 μg/ml; Undecane 290 μg/ml
Sertifiseringer: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.