Platina nominally 20% on carbon black

Leverandør: Thermo Fisher Scientific

47381.06 47381.03
47381.06EA 8325 NOK
47381.06 47381.03
Platina nominally 20% on carbon black
Platina
Formel: Pt
MDL nummer: MFCD00011179
CAS-nummer: 7440-06-4

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR