Reversed phase test mix 2, certified reference material, Supelco®
Leverandør: Merck
47641-U
47641-UEA 650 NOK
47641-U
Reversed phase test mix 2, certified reference material, Supelco®
Standarder kromatografi standarder
Analytes:
N,N-Diethyl-m-toluamide 600 μg/ml; Phenol 700 μg/ml; Toluene 4000 μg/ml; Uracil 5 μg/ml
Sertifiseringer: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Lagringstemp 2 - 8 °C