DL-α-Tocopherol acetate

Leverandør: Merck

Synonymer: (±)-α-Tocopherol acetate, all-rac-α-Tocopheryl acetate, Vitamine E acetate, DL-alpha-Tocopheryl acetate, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl acetate

47786
47786.EA 340 NOK
47786.
DL-α-Tocopherol acetate
DL-α-Tocopherol acetate
Formel: C₃₁H₅₂O₃
MW: 472.75 g/mol
MDL nummer: MFCD00072042
CAS-nummer: 7695-91-2
EINECS: 231-710-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9c92686d4071b9b984c6d75ad0fc374e33b5b67&vid=b9c92686d4071b9b984c6d75ad0fc374e33b5b67&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR