VWR®, Cryoboxes
Leverandør: VWR Collection
478-0241EA 37 NOK
478-0241
VWR®, Cryoboxes
Bokser Kryobokser
Polycarbonate.
  • Can be used from –190 to +121 °C
VWR® freezer boxes can be used with most manufacturer’s upright and chest freezer racks in general purpose and ultra-low mechanical freezers.