Dewar-kar, store, type C
Leverandør: KGW
1231 1232 1233 1234 1235 1236 1237 1241 1242 1243 1244 1245 1248 1253 1251 1255 1249 1258 1252
478-0734EA 7900 NOK
478-0734 478-0768 478-0769 478-0770 478-0771 478-0772 478-0773 478-4201 478-4202 478-4203 478-4204 478-4205 478-0774 478-0777 478-0776 478-0778 478-0775 478-0780 KGWA1252
Dewar-kar, store, type C
Dewars og kryogene fartøy
Dewar flasks made of borosilicate glass 3.3 DIN/ISO 3585, for storing and tranporting of LN₂, CO₂ and other cooling liquids. Suitable as insulating flasks.
  • With handle or two grips
  • Insulated lid (incl. grip)
  • The Dewar flasks are vacuum insulated and silvered
  • Large aperture
  • Robust design, with quick release clamp
Type C= metal - blue coated
Type CAL= Stucco aluminium
Temperature range: −200 to +200 °C.