Saccharin sodium hydrate

Leverandør: Merck

Synonymer: o-Benzoic sulphimide sodium salt hydrate, Sodium 3-oxo-3H-benzo[d]isothiazol-2-ide 1,1-dioxide hydrate, Saccharin, sodium salt hydrate

47839
47839.EA 340 NOK
47839.
Saccharin sodium hydrate
Saccharin sodium hydrate
Formel: C₇H₅NNaO₃S·H₂O
MW: 205.17 g/mol (anhy)
MDL nummer: MFCD00149605
CAS-nummer: 82385-42-0
EINECS: 204-886-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0ee6a8fb200fe46f483bfb2765a9c9d2db20cb9&vid=a0ee6a8fb200fe46f483bfb2765a9c9d2db20cb9&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR