1-Naphthyl phosphate monosodium salt monohydrate, Millipore®

Leverandør: Merck Millipore Calbiochem
Warning

479775-5
479775-5EA 1625 NOK
479775-5
1-Naphthyl phosphate monosodium salt monohydrate, Millipore®
1-Naphthyl phosphate monosodium salt monohydrate
Formel: C₁₀H₈NaO₄P·1H₂O
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00150615
CAS-nummer: 81012-89-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR