Autoclaves, LABOKLAV 25
Leverandør: SHP Steriltechnik
210025001000
481-0397EA 0 NOK
481-0397
Autoclaves, LABOKLAV 25
Sterilisatorer Autoklav, dampsterilisering
These compact benchtop units with a 25 litre useable chamber offers a large capacity and the performance of a big autoclave. The unit has a chamber door and a transparent front cover made from resistant safety glass, LED chamber illumination and coloured lighting to visualise the cycle progress. No feed water or waste water connection required, fan for fast cooling on some models, the user has only to plug it in and start. In stainless steel, chamber door made from heat-proof safety glass.
  • Integrated steam generator with dry heating elements, their