Connect Benchtop Autoclaves
Leverandør: CERTOCLAV
8510176
481-0824EA 0 NOK
481-0824
Connect Benchtop Autoclaves
Sterilisatorer Autoklav, dampsterilisering
Fully automatic laboratory autoclave for liquids sterilisation and documentation in the cloud. This autoclave has an 7" high-res touch screen and intuitive user interface All user tasks are explained with videos on the machine. Easy compliance with GLP documentation requirements possible via CertoCloud, where all user information and cycle data are safely stored for 10 years and can be accessed from any web browser (6 month free Certocloud trial period included with purchase).
  • Freely programmable autoclave with electrical lock and documentation in the