Classic Benchtop Autoclaves
Leverandør: CERTOCLAV
8510184 8510174
481-0826EA 49300 NOK
481-0826 481-0822
Classic Benchtop Autoclaves
Sterilisatorer Autoklav, dampsterilisering
The trusted, economic autoclaving solution for compact instruments also simply packaged, liquids (culture mediums), bottles and equipment, refuse sterilisation. For sterilisation with low requirement concerning documentation.
  • Process control via mechanical precision valve
  • Ultra-low maintenance autoclave with time tested technology
  • Interface: temperature selection knob
  • The simple operation allows for effortless work with less handling
  • Through the CertoLock locking device the laborious turning of several screws is avoided
  • 4-fold safety system
  • Manual time