VWR® VAPOUR-Line Lite II autoklaver
Leverandør: VWR Collection
481-0846EA 33625 NOK
481-0846 481-0847
VWR® VAPOUR-Line Lite II autoklaver
Sterilisatorer Autoklav, dampsterilisering
Autoclave for occasional use. AISI 304 stainless steel housing, easy to maintain and disinfect, aluminium inner chamber and ABS insulating lid with locking system.
  • Microprocessor-controlled automatic thermal regulation
  • Digital temperature and sterilisation time display
  • Automatic ventilation valve and electronic temperature cut-off safety protection system
  • Reinforced heating element and PT100 probe
  • Automatic sterilisation programs
  • Available with or without integrated printer
Instructions are GLP and EN 17205 compliant.
Bestillingsinformasjon: BasketMore Product Information