EDTA dinatrium salt concentrate for dilution to 0.01 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®

Leverandør: Avantor

Synonymer: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

4870.EA 475 NOK
4870.
EDTA dinatrium salt concentrate for dilution to 0.01 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
EDTA dinatrium salt
Formel: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00070672
CAS-nummer: 139-33-3
EINECS: 205-358-3
Merck Indeks: 12,03556

Bestill nå

Spesifikasjon Testresultater

Titer (mol/l) 0.0099-0.0101

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR