EDTA dinatrium salt concentrate for dilution to 0.1 mol/l in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Leverandør: Avantor
Synonymer: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt
Dose for preparation of 1 litre of solution.
4871.EA 585 NOK
4871.
EDTA dinatrium salt concentrate for dilution to 0.1 mol/l in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
EDTA dinatrium salt
Formel: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
MDL nummer: MFCD00070672
CAS-nummer: 139-33-3
Merck Indeks: 12,03556