Non-interfering Protein Assay™ kit, Calbiochem®

Leverandør: Merck Millipore Calbiochem

488250-1
488250-1EA 9950 NOK
488250-1
Non-interfering Protein Assay™ kit, Calbiochem®
Assay Proteinanalyser

Non-Interfering Protein Assay™ Kit is an easy-to-use protein assay that overcomes interference of agents found in protein solutions including detergents, chelating agents, reducing agents, amines, sugars, urea, and others. The Universal Protein Precipitating Agent (UPPA™ Reagent) is used to precipitate and immobilize the protein in the tube while the interfering reagents are removed.


  • Protein concentration is based on the specific binding of copper to the peptide backbone
  • With the increase in protein concentration, concentration of unbound copper ions decreases
  • Colour density is inversely related to the amount of protein present in solution


Leveringsinformasjon: This kit is supplied with UPPA™ reagents I and II, copper solution I, color agents A and B, BSA standard and a user protocol.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR