Surgical tape, Micropore™ 1530
Leverandør: 3M MEDICAL
M1530-0 M1530-1 M1530-2
115-8172EA 2900 NOK
115-8172 489-0024 489-0025
Surgical tape, Micropore™ 1530
Førstehjelpsutstyr Bandasjer
This hypoallergenic paper tape is gentle to the skin yet adheres well and leaves minimal adhesive residue upon removal.
  • Without natural rubber latex
  • Gentle adhesion
  • Conformable and highly breathable
  • Reliably secures dressings and devices to skin