Sterilisation indicators
Leverandør: SÜDPACK MEDICA
7600012
489-0046EA 670 NOK
489-0046
Sterilisation indicators
Steriliseringsindikatorer
Tapes indicators by dry heat sterilisation.
The tape (ethylene oxide) changes from white to black after 3,5 minutes at 134 °C.
Dry heat steriliser tape (poupinel) changes from green to different shades of brown after a period at a temperature between 155 and 180 °C.