Steam sterilisation indicators
Leverandør: SÜDPACK MEDICA
7600028
489-0100EA 3425 NOK
489-0100
Steam sterilisation indicators
Steriliseringsindikatorer
Steam sterilisation. The labels change colour from pink to dark brown after sterilisation.
Forpakning: Roll of 5000 labels.