Seal seam integrity test, Stericlin®
Leverandør: VP Medical Packaging
3FSZB830102 3FSZB830103 3FSZB830104
489-0144EA 1850 NOK
489-0144 489-0145 489-0146
Seal seam integrity test, Stericlin®
Lekkasjedetektorer Systemer for deteksjon av flytende lekkasje
Stericlin® seal seam integrity tests are used for routine testing of seal seams according to DIN EN ISO 11607. In combination with the Seal Test they offer comprehensive and secure routine testing for the heat sealing process.
  • Easy to perform
  • Quickly detects channels and voids
For specifications, see the DGSV guidelines for the validation of packaging processes according to DIN EN ISO 11607-2.
Bestillingsinformasjon: Seal seam integrity tests are available in two basic versions: 'Pipette' and 'disposable ink bag'.