2,4'-DDT

Leverandør: Merck
Danger

Synonymer: o,p'-DDT, 2,2,2,o,p'-Pentachloroethylidenebisbenzene

49018
49018.EA 3525 NOK
49018.
2,4'-DDT
2,4'-DDT
Formel: C₁₄H₉Cl₅
MW: 354.49 g/mol
CAS-nummer: 789-02-6
EINECS: 212-332-5
FN: 2761
ADR: 6.1 III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR