Multi-element standard solution 3 for ICP, TraceCERT®, Supelco®
Leverandør: Merck
49596-100ML
49596-100MLEA 1350 NOK
49596-100ML
Multi-element standard solution 3 for ICP, TraceCERT®, Supelco®
Standarder Kationstandarder Multi-Element Standards
Ca, 2000 mg/l
K, 200 mg/l
Mg, 400 mg/l
Na, 1000 mg/l
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034