Organic reference standard, Sclareol, analytical standard, Supelco®

Leverandør: Merck

49944-100MG
49944-100MGEA 855 NOK
49944-100MG
Organic reference standard, Sclareol, analytical standard, Supelco®
Standarder Organiske standarder
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a2d8f395977399b483095eb36f433174a30c1d2&vid=2a2d8f395977399b483095eb36f433174a30c1d2&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Beskrivelse Sclareol
Environmentally Preferable
CAS-nr 515-03-7
Formel C₂₀H₃₆O₂
Smeltepunkt 95 - 100 °C
Frysepunkt 218 - 220 °C/19 mmHg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR