Medium, Ham's F-12, Corning®
Leverandør: Corning
50-040-PB
50-040-PBEA 600 NOK
50-040-PB
Medium, Ham's F-12, Corning®
Media, Cellekultur
Ham's F-12 media was originally formulated for the serum-free culture of CHO cells, and is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids.
  • Low endotoxin - < 0,25 EU/ml
  • Mycoplasma tested
  • Liquid media formulations are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
This medium is also suitable for carcinoma cells, rat skeletal myoblasts, Chinese hamster lung cells, and rat, rabbit, and chicken embryos.