5,5'-difluoro BAPTA tetrapotassium salt ≥95% (by TLC) ion chelator, for calcium determination

Leverandør: Biotium

50006
50006.EA 4700 NOK
50006.
5,5'-difluoro BAPTA tetrapotassium salt ≥95% (by TLC) ion chelator, for calcium determination
5,5'-difluoro BAPTA tetrapotassium salt

BAPTA and its derivatives are calcium chelators that are commonly used to form calcium buffers with well-defined calcium concentrations. By injecting the chelators into cells or by incubating cells with the AM ester form of the chelators, one can control the cytosolic calcium concentration, an important means to study the roles of calcium.


  • Kd (no Mg2 ): 0.59 uM, Kd (1 mM Mg2 ): 0.70 uM
  • Cell permeability: Membrane impermeant
  • White solid soluble in water
  • Store at 4°C


Key advantages of these calcium chelators include relative insensitivity toward intracellular pH change and fast release of calcium. Biotium offers several BAPTA chelators with calcium dissociation constants covering the biologically significant range from 10-7 to 10-2 M.

Formel: C₂₂H₁₈F₂K₄N₂O₁₀
MW: 665 g/mol
Lagringstemperatur: Kjøleskap
CAS-nummer: 152290-47-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR