5,5’-Dimethyl BAPTA tetrapotassium salt ≥95% (by TLC) ion chelator, for calcium determination

Leverandør: Biotium

50008
50008.EA 2200 NOK
50008.
5,5’-Dimethyl BAPTA tetrapotassium salt ≥95% (by TLC) ion chelator, for calcium determination
5,5’-Dimethyl BAPTA tetrapotassium salt

BAPTA and its derivatives are calcium chelators that are commonly used to form calcium buffers with well-defined calcium concentrations. By injecting the chelators into cells or by incubating cells with the AM ester form of the chelators, one can control the cytosolic calcium concentration, an important means to study the roles of calcium.


  • Kd (no Mg2 ): 0.16 uM, Kd (1 mM Mg2 ): 0.44 uM
  • Cell permeability: Membrane impermeant
  • White solid soluble in water
  • Store at -20°C


Key advantages of these calcium chelators include relative insensitivity toward intracellular pH change and fast release of calcium. Biotium offers several BAPTA chelators with calcium dissociation constants covering the biologically significant range from 10-7 to 10-2 M.

Formel: C₂₄H₂₄K₄N₂O₁₀
MW: 657 g/mol
Lagringstemperatur: Fryseskap
CAS-nummer: 73630-09-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR