FLUO-3 pentasodium salt ≥95% (by HPLC) fluorescent indicator, for calcium determination

Leverandør: Biotium

50012
50012.EA 4125 NOK
50012.
FLUO-3 pentasodium salt ≥95% (by HPLC) fluorescent indicator, for calcium determination
FLUO-3 pentasodium salt

Fluo-3, pentasodium is membrane-impermeant, but can be loaded into cells via microinjection or scrape loading.


  • Kd: 450 nM
  • λEx/λEm (low or high [Ca2+]) = 506/526 nm
  • ε (506 nm) = 86,000 M-1cm-1
  • Cell permeability: Membrane impermeant
  • Orange red solid soluble in DMSO and water (pH >6)
  • Store at 4°C. Protect from light, especially when in solution

Formel: C₃₆H₂₅Cl₂Na₅N₂O₁₃
MW: 879 g/mol
Lagringstemperatur: Kjøleskap
CAS-nummer: 134907-84-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR