Rhod-590 AM ester fluorescent indicator, for calcium determination

Leverandør: Biotium

50025
50025.EA 7825 NOK
50025.
Rhod-590 AM ester fluorescent indicator, for calcium determination
Rhod-590 AM ester

Membrane-permeant form of Rhod-590 calcium indicator dye that can be loaded into cells via incubation. Because of the relatively low water solubility of the AM ester, Pluronic F-127, a mild detergent, is often used as a dispersing agent to facilitate dye loading. Rhod-590 AM ester itself does not bind Ca2 , but it is readily hydrolyzed to Rhod-590 by endogenous esterases once it enters cells.


  • Absorption/Emission 590nm/616nm (low [Ca2+] )
  • Absorption/Emission 595nm/626nm (high [Ca2+] )
  • Kd 0.61μM
  • Cell permeability: Membrane permeant
  • Dark purple solid soluble in DMSO
  • Store at -20°C and protect from light, especially in solution

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR