4-[(Dipropylamino)sulphonyl]benzoic acid sodium salt, water soluble

Leverandør: Biotium

Synonymer: Benemid sodium salt, Probenecid sodium salt

50027
50027.EA 4325 NOK
50027.
4-[(Dipropylamino)sulphonyl]benzoic acid sodium salt, water soluble
4-[(Dipropylamino)sulphonyl]benzoic acid sodium salt

Probenecid is commonly used to inhibit organic-anion transporters located in the cell membrane. Such transporters can extrude dyes and indicators and thus contribute to low signal or high background in assays due to poor cellular retention of dyes. The use of probenecid to block the efflux of intracellular dyes was first described by Di Virgilio et al. (1990), and it has been used with a wide range of anionic dyes and conjugates.


  • Colorless solid soluble in water (77mg/mL of water)


The commonly used free acid form of probenecid is difficult to dissolve, requiring 1M NaOH to prepare a solution. Our water-soluble probenecid sodium salt dissolves quickly in assay buffer and eliminates the need for handling caustic NaOH. Each vial is directly dissolved in 1 mL buffer to make a 250 mM stock solution. Typical working concentrations in cell-based assays are 1-2.5 mM.

Formel: C₁₃H₁₈NO₄SNa
MW: 307,37 g/mol
Lagringstemperatur: Romtemperatur
CAS-nummer: 23795-03-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR