Fura 2-AM ≥98% (by HPLC) fluorescent indicator, for calcium determination

Leverandør: Biotium

Synonymer: FURA-2 pentakis(acetoxymethyl) ester, 1-[2-(5-Carboxyoxazol-2-yl)-6-aminobenzofuran-5-oxy]-2-(2'-amino-5'-methyl-phenoxy)ethane-N,N,N',N'-tetraacetic acid, pentaacetoxymethyl ester, FURA-2 acetoxymethyl ester, Fura-2 AM Ester

50034 50033 50033-1
50034.EA 3600 NOK
50034. 50033. 50033-1
Fura 2-AM ≥98% (by HPLC) fluorescent indicator, for calcium determination
Fura 2-AM

Fura-2, AM ester is a ratiometric fluorescent dye that binds to free intracellular calcium, and is the preferred dye for ratiometric imaging microscopy. Fura-2, AM ester is membrane-permeant and thus can be loaded into cells via incubation.


  • Kd: 145 nM (22°C in pH 7.2 buffer) (after hydrolysis)
  • λEx/λEm (no Ca2+) = 363/512 nm; λEx/λEm (high Ca2+) = 335/505 nm (after hydrolysis)
  • ε (363 nm, no Ca2+) = 27,000 M-1cm-1; ε (335 nm, high Ca2+) = 35,000 M-1cm-1 (after hydrolysis)
  • Cell permeability: Membrane permeant
  • Light yellow solid soluble in DMSO
  • Store at -20°C and protect from light, especially in solution


Because of the relatively low water solubility of the AM ester, the mild detergent Pluronic F-127 (catalog no. 59004) is often used as a dispersing agent to facilitate cell loading. Fura-2, AM ester itself does not bind Ca2+, but it is readily hydrolyzed to Fura-2 by endogenous esterases once it enters cells.


Fura-2 fluorescence can be detected using flow cytometry, fluorescence microscopy, or certain fluoresence microplate readers. Upon calcium binding, the fluorescent excitation maximum of the indicator undergoes a blue shift from 363 nm (Ca2+-free) to 335 nm (Ca2+-saturated), while the fluorescence emission maximum is relatively unchanged at ~510 nm. A major use of Fura-2 is in drug discovery, for measuring intracellular calcium mobilization after activation of G protein-coupled receptors (GPCRs) and calcium channels.

Formel: C₄₄H₄₇N₃O₂₄
MW: 1001.857 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00036976
CAS-nummer: 108964-32-5

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR