Furaptra (Mag-Fura-2) AM Ester ≥95% (by TLC) fluorescent indicator, for calcium determination

Leverandør: Biotium

Synonymer: Mag-Fura-2 AM ester

50037 50039 50038
50037.EA 5750 NOK
50037. 50039. 50038.
Furaptra (Mag-Fura-2) AM Ester ≥95% (by TLC) fluorescent indicator, for calcium determination
Furaptra (Mag-Fura-2) AM Ester

Furaptra, AM ester is a membrane-permeant form of Furaptra (Mag-Fura-2). The indicator is loaded into cells via simple incubation


  • Absorption/Emission 366nm/475nm (EtOAc)
  • Extinction Coefficient 31,000
  • Cell permeability: Membrane permeant
  • Colors: Green
  • Light yellow solid soluble in DMSO
  • Store at -20°C and protect from light

Formel: C₃₀H₃₀N₂O₁₉
MW: 723 g/mol
Lagringstemperatur: Fryseskap
CAS-nummer: 130100-20-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR