HPLC columns, Discovery RP-AmideC16, Supelco®
Leverandør: Merck
505064-30 505013-40
505064-30EA 10060 NOK
505064-30 505013-40
HPLC columns, Discovery RP-AmideC16, Supelco®
Kromatografikolonner
These columns with excellent retention and resolution of polar compounds are less hydrophobic than C18 columns.
  • Unique selectivity compared to C18
  • Excellent peak shape and efficiency
  • Compatible with 100% aqueous mobile phases