Supelco® BIOshell™ HPLC Columns, Glycan Fused-Core®
Leverandør: Merck
50994-U
50994-UEA 12380 NOK
50994-U
Supelco® BIOshell™ HPLC Columns, Glycan Fused-Core®
Kromatografikolonner
BIOshell™ Glycan HPLC columns are specifically engineered to deliver fast, high resolution, reproducible gylcan identification using HILIC chromatography.
  • Fused-Core®particle provides a thin porous shell of high-purity silica surrounding a solid silica core
  • High column efficiency due to the shallow diffusion paths in the 0,5 µm thin porous shell
  • Small overall particle size of 2,7 µm
BIOshell™ Glycan columns consist of a proprietary, pentahydroxy chemistry tethered to a 2,7 µm, 90 Å superfivially porous prticle (SPP). Main application is for theMore Product Information