BCECF AM ester 95 HPLC_ASSAY_METHOD fluorescent pH indicator

Leverandør: Biotium

Synonymer: BCECF AM

51011 51011-1 51012
51011.EA 2675 NOK
51011. 51011-1 51012.
BCECF AM ester 95 HPLC_ASSAY_METHOD fluorescent pH indicator
BCECF AM ester

BCECF, AM ester is membrane-permeant and thus can be loaded into cells via incubation. This product is a mixture of three molecular species, all of which are readily hydrolyzed by endogenous esterases into a single BCECF free acid form once they are in the cells.


  • Cell permeability: Membrane permeant
  • Colors: Green
  • Absorption/Emission 508 nm/530 nm (for BCECF acid)
  • White solid soluble in DMSO and DMF
  • Store at -20°C and protect from light, especially in solution

Formel: C₄₂H₄₀O₂₁
MW: 808.69 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00036969
CAS-nummer: 117464-70-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=825a7550c456e5d203606af9dc6266ba39c0ff67&vid=9a4aca4f3bc9c03b7d450afef197f168ffbde83c&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR