5-Carboxyfluorescein 98 HPLC_ASSAY_METHOD fluorescent pH indicator

Leverandør: Biotium
Warning

Synonymer: Spiro(isobenzofuran-1(3H),9'-(9H)xanthene)-5-carboxylic acid, 3',6'-dihydroxy-3-oxo-, 5-FAM, 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylic acid, 5-Carboxyfluorescein hydrate

51019
51019.EA 2050 NOK
51019.
5-Carboxyfluorescein 98 HPLC_ASSAY_METHOD fluorescent pH indicator
5-Carboxyfluorescein

5-Carboxyfluorescein has a pKa of 6.5 and can be used as a pH sensor. The excitation spectrum of the dye responds to pH similarly to that of BCECF. Carboxyfluorescein can also be used as a tracer dye. The dye is membrane-impermeant and can be loaded into cells by microinjection or scrape loading.


  • λEx/λEm (pH 9) = 492/514 nm
  • pKa 6.5
  • Extinction Coefficient (ε) 82,000 (pH 9)
  • Yellow orange solid soluble in pH>6 water
  • Store at 4°C and protect from light, especially in solution

Formel: C₂₁H₁₂O₇.xH₂O
MW: 376.32 g/mol (anhy)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00036874
CAS-nummer: 76823-03-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05824cdba3ba0cda7a7d60efd2aeb0d37565def4&vid=572aefe2942bfeb12d0a638efa388e7a6b9130f1&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR