6-Carboxyfluorescein 98 HPLC_ASSAY_METHOD fluorescent pH indicator

Leverandør: Biotium
Warning

Synonymer: 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-6-carboxylic acid, 6-FAM

51020
51020.EA 2300 NOK
51020.
6-Carboxyfluorescein 98 HPLC_ASSAY_METHOD fluorescent pH indicator
6-Carboxyfluorescein

6-Carboxyfluorescein has a pKa of 6.5 and can be used as a pH sensor. The excitation spectrum of the dye responds to pH similarly to that of BCECF. Carboxyfluorescein can also be used as a tracer dye. The dye is membrane-impermeant and can be loaded into cells by microinjection or scrape loading.


  • λEx/λEm (pH 9) = 495/519 nm
  • pKa 6.5
  • Yellow orange solid soluble in water (pH>6), DMF or DMSO
  • Store at 4°C and protect from light

Formel: C₂₁H₁₂O₇
MW: 376.32 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00036873
CAS-nummer: 3301-79-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55462ecf1d90cd8fc0e3d006cd3bf5cf18695b6a&vid=93eb9da70e7f66fa5f5b624402beaa13ae182d22&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR