VWR®, Filter Holder, Stainless Steel
Leverandør: VWR Collection
511-0666EA 3675 NOK
511-0666
VWR®, Filter Holder, Stainless Steel
Filterholdere
Glass filter holder with 47 mm stainless steel screen, 300 ml glass funnel, 47 mm aluminium spring clamp, 47 mm glass base, PTFE gasket and silicone stopper no. 8.