Membrandispenser for mikrobiologisk kontroll, Membrane-Butler, Whatman™
Leverandør: Whatman products (Cytiva)
10477100
511-5500EA 32325 NOK
511-5500
Membrandispenser for mikrobiologisk kontroll, Membrane-Butler, Whatman™
Membrandispensere
Manual butler for dispensing filtration membranes.
  • High reliability
  • Cross contamination risks are minimized
  • Membrane dispensed rapidly
  • Highly suitable for use on sterile benches
  • Compact dimensions for portable use
The Membrane-Butler dispenses individual sterile MicroPlus and ME membrane filters (type 'STL'). The sterile packaging is inserted into the roller system and the system is ready to be used.
Membrane filters for microbiological checks must be handled carefully to ensure that they remain sterile and that quantitative results are beingMore Product Information