Centrifuge, Cellspin® III UNI
Leverandør: THARMAC GmbH
3200-220
512-0714EA 0 NOK
512-0714
Centrifuge, Cellspin® III UNI
Sentrifuger Sentrifuger, bordmodell
The genius among the centrifuges. Automatic rotor detection and speed adjustment on max. 2000 rpm when using cytorotors.
  • Huge rotor selection
  • No reprogramming required
  • Speed range: 200 to 15000 rpm
  • Sample volume: 0,1 to 5 ml (cytology), up to 4×200 ml (standard laboratory use)
Applicable rotors: swing out-, angle- as well as hematocrit-rotors and cyto-rotors, also compatible with Shandon™ Cytospin™ systems.
Sertifiseringer: CE and IVD certified.